Anti-NTRK3 Mouse Monoclonal Antibody [clone: 2B8]

Lieferant: OriGene
ORIGCF500537EA 749 EUR
ORIGCF500537
Anti-NTRK3 Mouse Monoclonal Antibody [clone: 2B8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NTRK3
Clonality: Monoclonal
Clone: 2B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR