Anti-SATB1 Mouse Monoclonal Antibody [clone: 6C6]

Lieferant: OriGene
ORIGCF500547EA 749 EUR
ORIGCF500547
Anti-SATB1 Mouse Monoclonal Antibody [clone: 6C6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SATB1
Clonality: Monoclonal
Clone: 6C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR