Anti-HSPA9 Mouse Monoclonal Antibody [clone: 4A1]

Lieferant: OriGene
ORIGCF500548EA 749 EUR
ORIGCF500548
Anti-HSPA9 Mouse Monoclonal Antibody [clone: 4A1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: HSPA9
Clonality: Monoclonal
Clone: 4A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR