Anti-C1orf161 Mouse Monoclonal Antibody [clone: 2A3]

Lieferant: OriGene
ORIGCF502507EA 770 EUR
ORIGCF502507
Anti-C1orf161 Mouse Monoclonal Antibody [clone: 2A3]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C1orf161
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR