Anti-ATP5B Mouse Monoclonal Antibody [clone: 1B10]

Lieferant: OriGene
ORIGCF500837EA 749 EUR
ORIGCF500837
Anti-ATP5B Mouse Monoclonal Antibody [clone: 1B10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: ATP5B
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR