Anti-UCK1 Mouse Monoclonal Antibody [clone: 4H1]

Lieferant: OriGene
ORIGCF501488EA 770 EUR
ORIGCF501488
Anti-UCK1 Mouse Monoclonal Antibody [clone: 4H1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: UCK1
Clonality: Monoclonal
Clone: 4H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR