Anti-AK3 Mouse Monoclonal Antibody [clone: 6B7]

Lieferant: OriGene
ORIGCF501835EA 749 EUR
ORIGCF501835
Anti-AK3 Mouse Monoclonal Antibody [clone: 6B7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: AK3
Clonality: Monoclonal
Clone: 6B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR