Anti-MICAL1 Mouse Monoclonal Antibody [clone: 6H10]

Lieferant: OriGene
ORIGCF501845EA 749 EUR
ORIGCF501845
Anti-MICAL1 Mouse Monoclonal Antibody [clone: 6H10]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100; Immunochemistry: 1:150

Type: Primary
Antigen: MICAL1
Clonality: Monoclonal
Clone: 6H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR