Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 1H6]

Lieferant: OriGene
ORIGCF501853EA 749 EUR
ORIGCF501853
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 1H6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ALDH1L1
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR