Anti-DUPD1 Mouse Monoclonal Antibody [clone: 1F3]

Lieferant: OriGene
ORIGCF501857EA 749 EUR
ORIGCF501857
Anti-DUPD1 Mouse Monoclonal Antibody [clone: 1F3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DUPD1
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR