Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 2A6]

Lieferant: OriGene
ORIGCF501869EA 749 EUR
ORIGCF501869
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 2A6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH1L1
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR