Anti-GAS7 Mouse Monoclonal Antibody [clone: 4E6]

Lieferant: OriGene
ORIGCF501872EA 770 EUR
ORIGCF501872
Anti-GAS7 Mouse Monoclonal Antibody [clone: 4E6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GAS7
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR