Anti-GBA3 Mouse Monoclonal Antibody [clone: 5G5]

Lieferant: OriGene
ORIGCF502533EA 749 EUR
ORIGCF502533
Anti-GBA3 Mouse Monoclonal Antibody [clone: 5G5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: GBA3
Clonality: Monoclonal
Clone: 5G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR