Anti-ANXA10 Mouse Monoclonal Antibody [clone: 4D8]

Lieferant: OriGene
ORIGCF500882EA 770 EUR
ORIGCF500882
Anti-ANXA10 Mouse Monoclonal Antibody [clone: 4D8]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ANXA10
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR