Anti-ELAVL1 Mouse Monoclonal Antibody [clone: 14F4]

Lieferant: OriGene
ORIGCF500887EA 749 EUR
ORIGCF500887
Anti-ELAVL1 Mouse Monoclonal Antibody [clone: 14F4]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: ELAVL1
Clonality: Monoclonal
Clone: 14F4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR