Anti-PLEK Mouse Monoclonal Antibody [clone: 6B5]

Lieferant: OriGene
ORIGCF500916EA 770 EUR
ORIGCF500916
Anti-PLEK Mouse Monoclonal Antibody [clone: 6B5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PLEK
Clonality: Monoclonal
Clone: 6B5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR