Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 6E3]

Lieferant: OriGene
ORIGCF501503EA 749 EUR
ORIGCF501503
Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 6E3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MOBKL2B
Clonality: Monoclonal
Clone: 6E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR