Anti-FAM84B Mouse Monoclonal Antibody [clone: 4A4]

Lieferant: OriGene
ORIGCF501916EA 749 EUR
ORIGCF501916
Anti-FAM84B Mouse Monoclonal Antibody [clone: 4A4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: FAM84B
Clonality: Monoclonal
Clone: 4A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR