Anti-UBXN2B Mouse Monoclonal Antibody [clone: 2E9]

Lieferant: OriGene
ORIGCF502560EA 770 EUR
ORIGCF502560
Anti-UBXN2B Mouse Monoclonal Antibody [clone: 2E9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: UBXN2B
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR