Anti-HOXC11 Mouse Monoclonal Antibody [clone: 4B8]

Lieferant: OriGene
ORIGCF502573EA 770 EUR
ORIGCF502573
Anti-HOXC11 Mouse Monoclonal Antibody [clone: 4B8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HOXC11
Clonality: Monoclonal
Clone: 4B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR