Anti-LACTB2 Mouse Monoclonal Antibody [clone: 3A5]

Lieferant: OriGene
ORIGCF502580EA 770 EUR
ORIGCF502580
Anti-LACTB2 Mouse Monoclonal Antibody [clone: 3A5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LACTB2
Clonality: Monoclonal
Clone: 3A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR