Anti-CISD1 Mouse Monoclonal Antibody [clone: 4G3]

Lieferant: OriGene
ORIGCF500927EA 749 EUR
ORIGCF500927
Anti-CISD1 Mouse Monoclonal Antibody [clone: 4G3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CISD1
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR