Anti-PSMC3 Mouse Monoclonal Antibody [clone: 7B10]

Lieferant: OriGene
ORIGCF500931EA 770 EUR
ORIGCF500931
Anti-PSMC3 Mouse Monoclonal Antibody [clone: 7B10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMC3
Clonality: Monoclonal
Clone: 7B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR