Anti-PSMC3 Mouse Monoclonal Antibody [clone: 10D12]

Lieferant: OriGene
ORIGCF500933EA 770 EUR
ORIGCF500933
Anti-PSMC3 Mouse Monoclonal Antibody [clone: 10D12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMC3
Clonality: Monoclonal
Clone: 10D12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR