Anti-CD80 Mouse Monoclonal Antibody [clone: 2B11]

Lieferant: OriGene
ORIGCF501575EA 749 EUR
ORIGCF501575
Anti-CD80 Mouse Monoclonal Antibody [clone: 2B11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR