Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 5A10]

Lieferant: OriGene
ORIGCF501940EA 749 EUR
ORIGCF501940
Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 5A10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: OSBPL11
Clonality: Monoclonal
Clone: 5A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR