Anti-OTUB2 Mouse Monoclonal Antibody [clone: 11B3]

Lieferant: OriGene
ORIGCF501946EA 749 EUR
ORIGCF501946
Anti-OTUB2 Mouse Monoclonal Antibody [clone: 11B3]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: OTUB2
Clonality: Monoclonal
Clone: 11B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR