Anti-PPWD1 Mouse Monoclonal Antibody [clone: 1G6]

Lieferant: OriGene
ORIGCF501948EA 770 EUR
ORIGCF501948
Anti-PPWD1 Mouse Monoclonal Antibody [clone: 1G6]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPWD1
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR