Anti-SULT1A1 Mouse Monoclonal Antibody [clone: 9B7]

Lieferant: OriGene
ORIGCF501951EA 749 EUR
ORIGCF501951
Anti-SULT1A1 Mouse Monoclonal Antibody [clone: 9B7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1A1
Clonality: Monoclonal
Clone: 9B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR