Anti-GBA3 Mouse Monoclonal Antibody [clone: 1C7]

Lieferant: OriGene
ORIGCF502604EA 749 EUR
ORIGCF502604
Anti-GBA3 Mouse Monoclonal Antibody [clone: 1C7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: GBA3
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR