Anti-LIPG Mouse Monoclonal Antibody [clone: 3G7]

Lieferant: OriGene
ORIGCF501000EA 749 EUR
ORIGCF501000
Anti-LIPG Mouse Monoclonal Antibody [clone: 3G7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: LIPG
Clonality: Monoclonal
Clone: 3G7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR