Anti-C16orf72 Mouse Monoclonal Antibody [clone: 3B4]

Lieferant: OriGene
ORIGCF501594EA 770 EUR
ORIGCF501594
Anti-C16orf72 Mouse Monoclonal Antibody [clone: 3B4]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: C16orf72
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR