Anti-CBWD1 Mouse Monoclonal Antibody [clone: 5H2]

Lieferant: OriGene
ORIGCF501611EA 770 EUR
ORIGCF501611
Anti-CBWD1 Mouse Monoclonal Antibody [clone: 5H2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CBWD1
Clonality: Monoclonal
Clone: 5H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR