Anti-ZIM2 Mouse Monoclonal Antibody [clone: 6D12]

Lieferant: OriGene
ORIGCF501975EA 770 EUR
ORIGCF501975
Anti-ZIM2 Mouse Monoclonal Antibody [clone: 6D12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: ZIM2
Clonality: Monoclonal
Clone: 6D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR