Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 7B3]

Lieferant: OriGene
ORIGCF501976EA 749 EUR
ORIGCF501976
Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 7B3]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500

Type: Primary
Antigen: OSBPL11
Clonality: Monoclonal
Clone: 7B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR