Anti-SPR Mouse Monoclonal Antibody [clone: 4F5]

Lieferant: OriGene
ORIGCF501983EA 770 EUR
ORIGCF501983
Anti-SPR Mouse Monoclonal Antibody [clone: 4F5]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SPR
Clonality: Monoclonal
Clone: 4F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR