Anti-PDLIM2 Mouse Monoclonal Antibody [clone: 9B6]

Lieferant: OriGene
ORIGCF502711EA 749 EUR
ORIGCF502711
Anti-PDLIM2 Mouse Monoclonal Antibody [clone: 9B6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDLIM2
Clonality: Monoclonal
Clone: 9B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR