Anti-SSX1 Mouse Monoclonal Antibody [clone: 2E9]

Lieferant: OriGene
ORIGCF502724EA 749 EUR
ORIGCF502724
Anti-SSX1 Mouse Monoclonal Antibody [clone: 2E9]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SSX1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR