Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1B3]

Lieferant: OriGene
ORIGCF501013EA 749 EUR
ORIGCF501013
Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1B3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: MTRF1L
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR