Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 6A6]

Lieferant: OriGene
ORIGCF501635EA 749 EUR
ORIGCF501635
Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 6A6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NSMCE2
Clonality: Monoclonal
Clone: 6A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR