Anti-PEPD Mouse Monoclonal Antibody [clone: 5D5]

Lieferant: OriGene
ORIGCF501655EA 770 EUR
ORIGCF501655
Anti-PEPD Mouse Monoclonal Antibody [clone: 5D5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PEPD
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR