Anti-RABL2A Mouse Monoclonal Antibody [clone: 4A3]

Lieferant: OriGene
ORIGCF502019EA 749 EUR
ORIGCF502019
Anti-RABL2A Mouse Monoclonal Antibody [clone: 4A3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RABL2A
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR