Anti-SNTA1 Mouse Monoclonal Antibody [clone: 2D5]

Lieferant: OriGene
ORIGCF502041EA 749 EUR
ORIGCF502041
Anti-SNTA1 Mouse Monoclonal Antibody [clone: 2D5]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNTA1
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR