Anti-FGFR2 Mouse Monoclonal Antibody [clone: 1D3]

Lieferant: OriGene
ORIGCF502797EA 749 EUR
ORIGCF502797
Anti-FGFR2 Mouse Monoclonal Antibody [clone: 1D3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FGFR2
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR