Anti-PPM1B Mouse Monoclonal Antibody [clone: 3C2]

Lieferant: OriGene
ORIGCF502812EA 749 EUR
ORIGCF502812
Anti-PPM1B Mouse Monoclonal Antibody [clone: 3C2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PPM1B
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR