Anti-PIM2 Mouse Monoclonal Antibody [clone: 8G10]

Lieferant: OriGene
ORIGCF501060EA 770 EUR
ORIGCF501060
Anti-PIM2 Mouse Monoclonal Antibody [clone: 8G10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PIM2
Clonality: Monoclonal
Clone: 8G10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR