Anti-PIM2 Mouse Monoclonal Antibody [clone: 5D5]

Lieferant: OriGene
ORIGCF501063EA 770 EUR
ORIGCF501063
Anti-PIM2 Mouse Monoclonal Antibody [clone: 5D5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: PIM2
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR