Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1F11]

Lieferant: OriGene
ORIGCF501065EA 749 EUR
ORIGCF501065
Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1F11]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: MTRF1L
Clonality: Monoclonal
Clone: 1F11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR