Anti-MTRF1L Mouse Monoclonal Antibody [clone: 2B5]

Lieferant: OriGene
ORIGCF501067EA 749 EUR
ORIGCF501067
Anti-MTRF1L Mouse Monoclonal Antibody [clone: 2B5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MTRF1L
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR