Anti-DNAJA2 Mouse Monoclonal Antibody [clone: 1C4]

Lieferant: OriGene
ORIGCF501688EA 749 EUR
ORIGCF501688
Anti-DNAJA2 Mouse Monoclonal Antibody [clone: 1C4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJA2
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR